Skip to Content
Merck
All Photos(1)

Key Documents

726796

Sigma-Aldrich

(S)-(+)-1-Cyclohexylethylamine

ChiPros®, produced by BASF, 99%

Synonym(s):

(S)-(+)-α-Methylcyclohexanemethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CH(CH3)NH2
CAS Number:
Molecular Weight:
127.23
Beilstein:
2935069
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

grade

produced by BASF

Quality Level

Assay

≥98.5% (GC)
99%

form

liquid

optical purity

enantiomeric excess: ≥98.5%

refractive index

n20/D 1.4614 (lit.)

bp

60 °C/12 mmHg (lit.)

density

0.856 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

C[C@H](N)C1CCCCC1

InChI

1S/C8H17N/c1-7(9)8-5-3-2-4-6-8/h7-8H,2-6,9H2,1H3/t7-/m0/s1

InChI key

XBWOPGDJMAJJDG-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(S)-(+)-1-Cyclohexylethylamine is a chiral amine.

Application

(S)-(+)-1-Cyclohexylethylamine may be used in the synthesis of (S)-(-)-ferrocenylimine by reacting with ferrocenecarboxaldehyde.

Legal Information

ChiPros is a registered trademark of BASF SE

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isoindolinones via a room temperature palladium nanoparticle-catalysed 3-component cyclative carbonylation?amination cascade
Grigg, Ronald, et al.
Tetrahedron, 44.37, 6979-6982 (2003)
Yee Voan Teo et al.
Cell reports, 27(4), 997-1007 (2019-04-25)
Oncogene-induced senescence (OIS) is a tumor suppressive response to oncogene activation that can be transmitted to neighboring cells through secreted factors of the senescence-associated secretory phenotype (SASP). Currently, primary and secondary senescent cells are not considered functionally distinct endpoints. Using
Supramolecular chirogenesis in bis (zinc porphyrin): an absolute configuration probe highly sensitive to guest structure
Borovkov, Victor V., Juha M. Lintuluoto, and Yoshihisa Inoue
Organic Letters, 2.11, 1565-1568 (2000)
Expedient synthesis and design strategies for new peptoid construction
Gorske, Benjamin C., et al.
Organic Letters, 7.8, 1521-1524 (2005)
Synthesis of optically-active planar chiral derivatives of ferrocene. Crystal structures of alkyne insertion products.
Zhao G, et al.
Tetrahedron Asymmetry, 9(13), 2253-2257 (1998)

Articles

Chiral amines play an important role in stereoselective organic synthesis. They are used directly as resolving agents, building blocks, or chiral auxiliaries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service