Recommended Products
Quality Level
Assay
95%
form
solid
mp
114-118 °C
SMILES string
ClC(=O)c1nc2ccccc2s1
InChI
1S/C8H4ClNOS/c9-7(11)8-10-5-3-1-2-4-6(5)12-8/h1-4H
InChI key
AOIGQLLPWDXVGB-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service