Skip to Content
Merck
All Photos(1)

Documents

700762

Sigma-Aldrich

(S)-SIPHOS-PE

97%

Synonym(s):

(11aS)-(−)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1′, 7′-fg][1,3,2]dioxaphosphocin-5-bis[(R)-1-phenylethyl]amine, N-Di[(R)-1-phenylethyl]-[(S)-1,1′-spirobiindane-7,7′-diyl]-phosphoramidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H32NO2P
CAS Number:
Molecular Weight:
505.59
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

optical activity

[α]22/D −65.0°, c = 1 in chloroform

mp

120-125 °C

storage temp.

−20°C

SMILES string

C[C@@H](N([C@H](C)c1ccccc1)P2Oc3cccc4CCC5(CCc6cccc(O2)c56)c34)c7ccccc7

InChI

1S/C33H32NO2P/c1-23(25-11-5-3-6-12-25)34(24(2)26-13-7-4-8-14-26)37-35-29-17-9-15-27-19-21-33(31(27)29)22-20-28-16-10-18-30(36-37)32(28)33/h3-18,23-24H,19-22H2,1-2H3/t23-,24-,33?/m1/s1

InChI key

ZXLQLIDJAURBDD-HRPAVAKOSA-N

Application

(S)-SIPHOS-PE is a chiral spiro phosphoramidate ligand that can be used to synthesize:
  • Spiroindolenines by Pd-catalyzed intramolecular dearomative arylation of haloaryl-3-substituted indoles.
  • Enantioselective conjugate addition compound by Cu-catalyzed 1,4-addition of diethylzinc (Et2Zn) to cyclic enones.
  • Octaketide natural products via rhodium-phosphoramidite catalyzed alkene hydroacylation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly enantioselective copper-catalyzed conjugate addition of diethylzinc to enones using chiral spiro phosphoramidites as ligands
Zhou H, et al.
The Journal of Organic Chemistry, 68(4), 1582-1584 (2003)
Rhodium-phosphoramidite catalyzed alkene hydroacylation: Mechanism and octaketide natural product synthesis
von Delius M, et al.
Journal of the American Chemical Society, 134(36), 15022-15032 (2012)
Palladium (0)-catalyzed dearomative arylation of indoles: convenient access to spiroindolenine derivatives
Wu K-J, et al.
Organic Letters, 14(14), 3772-3775 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service