Skip to Content
Merck
All Photos(1)

Documents

697877

Sigma-Aldrich

Tetramethyl(n-propyl)cyclopentadiene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20
CAS Number:
Molecular Weight:
164.29
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

reaction suitability

reagent type: ligand

refractive index

n20/D 1.474

density

0.811 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCCC1C(C)=C(C)C(C)=C1C

InChI

1S/C12H20/c1-6-7-12-10(4)8(2)9(3)11(12)5/h12H,6-7H2,1-5H3

InChI key

SQGUQDKNIMNMLY-UHFFFAOYSA-N

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

160.0 °F

Flash Point(C)

71.1 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service