Skip to Content
Merck
All Photos(1)

Key Documents

672858

Sigma-Aldrich

4-(Trifluoromethoxy)phenylacetylene

97%

Synonym(s):

4-Ethynyl-1-(trifluoromethoxy)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5F3O
CAS Number:
Molecular Weight:
186.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.4554

density

1.215 g/mL at 25 °C

functional group

fluoro

SMILES string

FC(F)(F)Oc1ccc(cc1)C#C

InChI

1S/C9H5F3O/c1-2-7-3-5-8(6-4-7)13-9(10,11)12/h1,3-6H

InChI key

RWWGGRCLMVYXPM-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F

Flash Point(C)

40 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service