Skip to Content
Merck
All Photos(1)

Key Documents

660426

Sigma-Aldrich

Gold nanoparticles

2-4 nm avg. part. size (TEM), octanethiol functionalized, 2 % (w/v) in toluene

Synonym(s):

Octylthiol functionalized gold nanoparticles

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

description

nanoparticles

Quality Level

form

nanoparticles
solution

concentration

2 % (w/v) in toluene

avg. part. size

2-4 nm (TEM)

density

0.8737 g/mL at 25 °C

functional group

thiol

storage temp.

2-8°C

SMILES string

CCCCCCCCS[Au]

InChI

1S/C8H18S.Au/c1-2-3-4-5-6-7-8-9;/h9H,2-8H2,1H3;/q;+1/p-1

InChI key

NXXPCKKHXAOGCX-UHFFFAOYSA-M

Features and Benefits

Useful for the self-assembly of gold structures for nanoelectronics.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

44.6 °F - closed cup

Flash Point(C)

7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service