Skip to Content
Merck
All Photos(1)

Key Documents

614343

Sigma-Aldrich

4-Nonylphenol-2,3,5,6-d4

97 atom % D, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15D4H20O
CAS Number:
Molecular Weight:
224.38
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

97 atom % D

Assay

98% (CP)

form

solid

bp

293-297 (lit.)

density

0.957 g/mL

mass shift

M+4

SMILES string

[2H]c1c([2H])c(CCCCCCCCC)c([2H])c([2H])c1O

InChI

1S/C15H24O/c1-2-3-4-5-6-7-8-9-14-10-12-15(16)13-11-14/h10-13,16H,2-9H2,1H3/i10D,11D,12D,13D

InChI key

IGFHQQFPSIBGKE-ZGAVCIBUSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilaria Di Marco Pisciottano et al.
Food additives & contaminants. Part B, Surveillance, 13(2), 139-147 (2020-03-27)
An analytical method is applied to simultaneously determine bisphenol A, 4-octylphenol, 4-tert-octylphenol, 4-nonylphenol and tert-nonylphenol in fish muscle. The method consisted of a cleanup on molecularly imprinted polymers cartridges (MIPs) followed by liquid chromatography coupled to a tandem mass spectrometer.
Ghada Aborkhees et al.
Molecules (Basel, Switzerland), 25(17) (2020-09-02)
A liquid chromatography-negative ion electrospray ionization-tandem mass spectrometry method was developed for the simultaneous analysis of bisphenol A, 4-octylphenol, 4-nonylphenol, diethylstilbestrol, 17β-estradiol, estriol, estrone, 17α-ethinylestradiol, prednisone, and prednisolone. This method used solid-phase extraction with an elution solvent of acetonitrile to
Nicoletta Ademollo et al.
Marine pollution bulletin, 163, 111900-111900 (2020-12-20)
This work aimed to investigate the contamination pattern in Kongsfjorden marine environment (Svalbard, 79°N 12°E) and to disentangle primary and secondary emissions. Surface seawater, sampled in two seasons, was analysed by GC-MS and LC-MS/MS to detect polychlorobiphenyls (PCBs), polycyclic aromatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service