596604
Aniline-13C6 hydrochloride
99 atom % 13C
Synonym(s):
Benzenamine-1,2,3,4,5,6-13C6, hydrochloride
About This Item
isotopic purity
99 atom % 13C
Assay
99% (CP)
mass shift
M+6
storage temp.
2-8°C
SMILES string
Cl.N[13c]1[13cH][13cH][13cH][13cH][13cH]1
InChI
1S/C6H7N.ClH/c7-6-4-2-1-3-5-6;/h1-5H,7H2;1H/i1+1,2+1,3+1,4+1,5+1,6+1;
InChI key
MMCPOSDMTGQNKG-BVNCJLROSA-N
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1 - STOT RE 1
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
381.2 °F - closed cup
Flash Point(C)
194 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service