Skip to Content
Merck
All Photos(1)

Key Documents

589888

Sigma-Aldrich

Methylamine-d5

99 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3ND2
CAS Number:
Molecular Weight:
36.09
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

99 atom % D

Quality Level

bp

-6.3 °C (lit.)

mass shift

M+5

SMILES string

[2H]N([2H])C([2H])([2H])[2H]

InChI

1S/CH5N/c1-2/h2H2,1H3/i1D3/hD2

InChI key

BAVYZALUXZFZLV-OMNVKBNWSA-N

Packaging

Standard 10 g quantity is packaged in a 450 mL carbon steel lecture bottle with stainless steel CGA 110/180 valve. Nominal gas pressure at 21ºC is 29 psig for 10 g. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Flam. Gas 1A - Press. Gas Liquefied gas - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service