Skip to Content
Merck
All Photos(1)

Key Documents

586633

Sigma-Aldrich

Acrylonitrile-2-13C

≥99 atom % 13C, ≥99% (CP), contains hydroquinone as stabilizer

Synonym(s):

2-Propenenitrile-2-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=13CHCN
CAS Number:
Molecular Weight:
54.06
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

≥99 atom % 13C

Assay

≥99% (CP)

contains

hydroquinone as stabilizer

bp

77 °C (lit.)

mp

-83 °C (lit.)

density

0.830 g/mL at 25 °C

mass shift

M+1

storage temp.

2-8°C

SMILES string

C=[13CH]C#N

InChI

1S/C3H3N/c1-2-3-4/h2H,1H2/i2+1

InChI key

NLHHRLWOUZZQLW-VQEHIDDOSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

23.0 °F - closed cup

Flash Point(C)

-4.99 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service