541575
2,4-Diamino-6-(hydroxymethyl)pteridine hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H8N6O ·HCl
CAS Number:
Molecular Weight:
228.64
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
mp
220 °C (lit.)
functional group
hydroxyl
SMILES string
Cl[H].Nc1nc(N)c2nc(CO)cnc2n1
InChI
1S/C7H8N6O.ClH/c8-5-4-6(13-7(9)12-5)10-1-3(2-14)11-4;/h1,14H,2H2,(H4,8,9,10,12,13);1H
InChI key
XZHMPUJCSYVIQL-UHFFFAOYSA-N
Related Categories
Application
2,4-Diamino-6-(hydroxymethyl)pteridine hydrochloride may be used in the synthesis of 6-bromomethyl-pteridine-2,4-diamine.
Reactant involved in the synthesis of prodrugs
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Genbin Shi et al.
Bioorganic & medicinal chemistry, 20(1), 47-57 (2011-12-16)
6-Hydroxymethyl-7,8-dihydropterin pyrophosphokinase (HPPK), a key enzyme in the folate biosynthetic pathway, catalyzes the pyrophosphoryl transfer from ATP to 6-hydroxymethyl-7,8-dihydropterin. The enzyme is essential for microorganisms, is absent from humans, and is not the target for any existing antibiotics. Therefore, HPPK
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service