Skip to Content
Merck
All Photos(1)

Documents

498998

Sigma-Aldrich

4-Methyl-2-pyridylzinc bromide solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6BrNZn
CAS Number:
Molecular Weight:
237.41
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

concentration

0.5 M in THF

density

0.985 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Cc1ccnc([Zn]Br)c1

InChI

1S/C6H6N.BrH.Zn/c1-6-2-4-7-5-3-6;;/h2-4H,1H3;1H;/q;;+1/p-1

InChI key

AHMFWLQZMAULAV-UHFFFAOYSA-M

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3 - Water-react 2

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service