Skip to Content
Merck
All Photos(1)

Key Documents

492310

Sigma-Aldrich

3-Hydroxytetradecanoic acid-2,2,3,4,4-d5

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9CD2CD(OH)CD2CO2H
CAS Number:
Molecular Weight:
249.40
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mp

78-80 °C (lit.)

mass shift

M+5

SMILES string

[2H]C([2H])(CCCCCCCCCC)C([2H])(O)C([2H])([2H])C(O)=O

InChI

1S/C14H28O3/c1-2-3-4-5-6-7-8-9-10-11-13(15)12-14(16)17/h13,15H,2-12H2,1H3,(H,16,17)/i11D2,12D2,13D

InChI key

ATRNZOYKSNPPBF-RWKHAKRRSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Del Bas et al.
International journal of obesity (2005), 42(4), 746-754 (2017-11-24)
Gut microbiota have been described as key factors in the pathophysiology of obesity and different components of metabolic syndrome (MetS). The cafeteria diet (CAF)-fed rat is a preclinical model that reproduces most of the alterations found in human MetS by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service