Skip to Content
Merck
All Photos(3)

Key Documents

423769

Sigma-Aldrich

α-Vinylbenzyl alcohol

97%

Synonym(s):

α-Ethenylbenzenemethanol, α-Phenylallyl alcohol, (±)-1-Phenylallyl alcohol, 1-Hydroxy-1-phenyl-2-propene, 1-Phenyl-2-propen-1-ol, 1′-Hydroxyallylbenzene, 3-Phenyl-3-hydroxy-1-propene, Phenylvinylcarbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH=CH2)OH
CAS Number:
Molecular Weight:
134.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.54 (lit.)

bp

101-102 °C/12 mmHg (lit.)

density

1.021 g/mL at 25 °C (lit.)

functional group

hydroxyl
phenyl

SMILES string

OC(C=C)c1ccccc1

InChI

1S/C9H10O/c1-2-9(10)8-6-4-3-5-7-8/h2-7,9-10H,1H2

InChI key

MHHJQVRGRPHIMR-UHFFFAOYSA-N

General description

α-Vinylbenzyl alcohol is an allyl alcohol derivative. Its isomerization reaction using a novel ternary catalyst system has been reported. The kinetic resolution of racemic α-vinylbenzyl alcohol by tungsten-catalyzed asymmetric epoxidation method has been repoorted. The coupling reaction of α-vinylbenzyl alcohol and aldehydes in the presence of RuCl2(PPh3)3 catalyst and In(OAc)3 as a co-catalyst in ionic liquids has been investigated. Etherification reaction of glycerol with α-vinylbenzyl alcohol in the presence of sulfonic catalyst immobilized on silica (SiO2–SO3H) has been reported.

Application

α−Vinylbenzyl alcohol may be used in the synthesis of cinnamyl ethers by reacting with alcohols.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heterogeneously catalyzed etherification of glycerol: new pathways for transformation of glycerol to more valuable chemicals.
Gu Y, et al.
Green Chemistry, 10(2), 164-167 (2008)
Masato Ito et al.
Journal of the American Chemical Society, 127(17), 6172-6173 (2005-04-28)
Highly efficient isomerization of allylic alcohols into saturated carbonyls is accomplished using the catalyst system of Cp*RuCl[Ph2P(CH2)2NH2-kappa2-P,N]-KOt-Bu (Cp* = eta5-C5(CH3)5) under mild conditions. Mechanistic consideration based on isotope-labeling experiments indicated the present reaction is applicable to the asymmetric isomerization of
Chuan Wang et al.
Journal of the American Chemical Society, 136(4), 1222-1225 (2014-01-16)
A simple, efficient, and environmentally friendly asymmetric epoxidation of primary, secondary, tertiary allylic, and homoallylic alcohols has been accomplished. This process was promoted by a tungsten-bishydroxamic acid complex at room temperature with the use of aqueous 30% H2O2 as oxidant
Ruthenium-catalyzed tandem olefin migration-Aldol-and mannich-type reactions in ionic liquid.
Yang XF, et al.
J. Mol. Catal. A: Chem., 214(1), 147-154 (2004)
Yoshio Kasashima et al.
Journal of oleo science, 59(10), 549-555 (2010-09-30)
Reactions of α-vinylbenzyl alcohol with other alcohols using iodine as a catalyst were investigated. The corresponding cinnamyl ethers were obtained as products. This suggested that α-vinylbenzyl alcohol was converted to cinnamyl ethers via 1-phenylallyl cation. Cinnamyl ethyl ether was obtained

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service