Skip to Content
Merck
All Photos(1)

Documents

412090

Sigma-Aldrich

Bisphenol A ethoxylate diacrylate

average Mn ~512, EO/phenol 2

Synonym(s):

Bis(4-acryloxypolyethoxyphenyl)propane, Bisphenol A polyethylene glycol diether diacrylate, Ethoxylated bisphenol A diacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[H2C=CHCO2(CH2CH2O)nC6H4-4-]2C(CH3)2
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

viscous liquid

Quality Level

mol wt

average Mn ~512

contains

1,000 ppm monomethyl ether hydroquinone as inhibitor

composition

EO/phenol, 2

refractive index

n20/D 1.534

density

1.14 g/mL at 25 °C

SMILES string

OCCO.OC(=O)C=C.CC(C)(c1ccc(O)cc1)c2ccc(O)cc2

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service