Skip to Content
Merck
All Photos(1)

Key Documents

332623

Sigma-Aldrich

Triacetyl-β-cyclodextrin

Synonym(s):

TAβCD, triacetyl-β-CD, β-Cyclodextrin heneicosaacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C84H112O56
CAS Number:
Molecular Weight:
2017.75
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

form

powder

optical activity

[α]25/D +125°, c = 1 in chloroform

mp

204-206 °C (lit.)

SMILES string

CC(=O)OC[C@H]1O[C@@H]2O[C@@H]3[C@@H](COC(C)=O)O[C@H](O[C@@H]4[C@@H](COC(C)=O)O[C@H](O[C@@H]5[C@@H](COC(C)=O)O[C@H](O[C@@H]6[C@@H](COC(C)=O)O[C@H](O[C@@H]7[C@@H](COC(C)=O)O[C@H](O[C@@H]8[C@@H](COC(C)=O)O[C@H](O[C@H]1[C@@H](OC(C)=O)[C@@H]2OC(C)=O)[C@@H](OC(C)=O)[C@@H]8OC(C)=O)[C@@H](OC(C)=O)[C@@H]7OC(C)=O)[C@@H](OC(C)=O)[C@@H]6OC(C)=O)[C@@H](OC(C)=O)[C@@H]5OC(C)=O)[C@@H](OC(C)=O)[C@@H]4OC(C)=O)[C@@H](OC(C)=O)[C@@H]3OC(C)=O

InChI

1S/C84H112O56/c1-29(85)106-22-50-57-64(113-36(8)92)71(120-43(15)99)78(127-50)135-58-51(23-107-30(2)86)129-80(73(122-45(17)101)65(58)114-37(9)93)137-60-53(25-109-32(4)88)131-82(75(124-47(19)103)67(60)116-39(11)95)139-62-55(27-111-34(6)90)133-84(77(126-49(21)105)69(62)118-41(13)97)140-63-56(28-112-35(7)91)132-83(76(125-48(20)104)70(63)119-42(14)98)138-61-54(26-110-33(5)89)130-81(74(123-46(18)102)68(61)117-40(12)96)136-59-52(24-108-31(3)87)128-79(134-57)72(121-44(16)100)66(59)115-38(10)94/h50-84H,22-28H2,1-21H3/t50-,51-,52-,53-,54-,55-,56-,57-,58-,59-,60-,61-,62-,63-,64-,65-,66-,67-,68-,69-,70-,71+,72+,73+,74+,75+,76+,77?,78-,79-,80-,81-,82-,83-,84-/m1/s1

InChI key

NOPKOJDDVCBPTP-URXASQAJSA-N

General description

Triacetyl-β-cyclodextrin, a water-insoluble derivative of cyclodextrin typically formed by the substitution of a hydroxyl group of the parent compound with maleic anhydride (−OCOCH3), has been proposed to act as a sustained release carrier for some pharmaceutical compounds.

Application

Invovled in studies of:
  • Controlled drug release of an inclusion complex
  • Biocompatible excipients for solubilization and deaggregation of cobalt bis(dicarbollide) derivatives in water
  • Extraction of aromatic amino acids
  • Emulsifiers
  • Polymerization initiation of cyclic esters
  • Sustained release matrix tablets

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J H Luong et al.
Journal of chromatography. A, 864(2), 323-333 (2000-02-11)
Nonaqueous capillary electrophoresis (NACE) equipped with amperometric detection has been developed for separation and detection of an 11-member model mixture of chlorinated phenolic compounds. With triacetyl-beta-cyclodextrin (TACD) as a novel selectivity selector, acetonitrile proved to be an excellent solvent for
Eighteenth IAPRI World Packaging Conference
Eighteen Steps to a Healthy Life (2012)
Catarina Marques Fernandes et al.
Chemical & pharmaceutical bulletin, 50(12), 1597-1602 (2002-12-25)
The inclusion ability of triacetyl-beta-cyclodextrin (TAbetaCD), a hydrophobic cyclodextrin (CD) derivative was examined, using nicardipine hydrochloride (NC) as model drug. The binary compounds were prepared in a 1 : 1 molar ratio by the kneading and the spray-drying techniques. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service