All Photos(1)
About This Item
Linear Formula:
ClCS2C6H5
CAS Number:
Molecular Weight:
188.70
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
95%
form
liquid
refractive index
n20/D 1.6688 (lit.)
bp
135 °C/15 mmHg (lit.)
density
1.331 g/mL at 25 °C (lit.)
functional group
chloro
thioether
storage temp.
2-8°C
SMILES string
ClC(=S)Sc1ccccc1
InChI
1S/C7H5ClS2/c8-7(9)10-6-4-2-1-3-5-6/h1-5H
InChI key
XEXIHCWFFNGQDE-UHFFFAOYSA-N
General description
Kinetics and mechanism of solvolysis reaction of phenyl chlorodithioformate was reported.
Application
Phenyl chlorodithioformate was used in synthesis of phenyl arylsulfonyl-alkyl-dithiocarbamates. It was also used in synthesis of S-phenyl O-(4-nitrophenyl)dithiocarbonate.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Kinetics and mechanism of the reactions of aryl chlorodithioformates with pyridines and secondary alicyclic amines.
Castro EA, et al.
Journal of Physical Organic Chemistry, 22(11), 1030-1037 (2009)
Synthesis of Phenyl Arylsulfonyl-alkyl-dithiocarbamates and Their Hydrolytic Reactivity in Hydroxide and Hydroperoxide Media.
Norberto F, et al.
European Journal of Organic Chemistry, 2005(21), 4710-4714 (2005)
The nucleofuge in the pyridinolysis of O-(4-nitrophenyl) S-aryl thio and dithiocarbonates.
Castro EA, et al.
Journal of Physical Organic Chemistry, 25(11), 994-994 (2012)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service