Skip to Content
Merck
All Photos(1)

Key Documents

30681

Sigma-Aldrich

L-4-Hydroxyproline methyl ester hydrochloride

≥98.0% (GC)

Synonym(s):

(2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid methyl ester hydrochloride, trans-4-Hydroxyproline methyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO3 · HCl
CAS Number:
Molecular Weight:
181.62
Beilstein:
4716932
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

SMILES string

Cl.COC(=O)[C@@H]1C[C@@H](O)CN1

InChI

1S/C6H11NO3.ClH/c1-10-6(9)5-2-4(8)3-7-5;/h4-5,7-8H,2-3H2,1H3;1H/t4-,5+;/m1./s1

InChI key

KLGSHNXEUZOKHH-JBUOLDKXSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily M Christensen et al.
The Journal of biological chemistry, 295(52), 18316-18327 (2020-10-29)
Pyrroline-5-carboxylate reductase 1 (PYCR1) catalyzes the biosynthetic half-reaction of the proline cycle by reducing Δ1-pyrroline-5-carboxylate (P5C) to proline through the oxidation of NAD(P)H. Many cancers alter their proline metabolism by up-regulating the proline cycle and proline biosynthesis, and knockdowns of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service