Skip to Content
Merck
All Photos(1)

Key Documents

296759

Sigma-Aldrich

Iodomethane-12C,d3

99.9 atom % 12C, 98 atom % D, 99% (CP), contains copper as stabilizer

Synonym(s):

Methyl-12C,d3 iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
12CD3I
CAS Number:
Molecular Weight:
144.95
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

vapor pressure

22.83 psi ( 55 °C)
6.66 psi ( 20 °C)

description

13C-depleted

isotopic purity

99.9 atom % 12C
98 atom % D

Assay

99% (CP)

contains

copper as stabilizer

bp

42 °C (lit.)

mp

-66.5 °C (lit.)

density

2.330 g/mL at 25 °C

mass shift

M+3

storage temp.

2-8°C

SMILES string

I[12C]([2H])([2H])[2H]

InChI

1S/CH3I/c1-2/h1H3/i1+0D3

InChI key

INQOMBQAUSQDDS-GIOFAXNUSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service