Skip to Content
Merck
All Photos(3)

Key Documents

241571

Sigma-Aldrich

Quinoline

reagent grade, 98%

Synonym(s):

1-Benzazine, 2,3-Benzopyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7N
CAS Number:
Molecular Weight:
129.16
Beilstein:
107477
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

reagent grade

vapor density

4.5 (vs air)

vapor pressure

0.07 mmHg ( 20 °C)

Assay

98%

form

liquid

autoignition temp.

896 °F

refractive index

n20/D 1.625 (lit.)

bp

113-114 °C/11 mmHg (lit.)
237 °C (lit.)

mp

−17-−13 °C (lit.)

density

1.093 g/mL at 25 °C (lit.)

SMILES string

c1ccc2ncccc2c1

InChI

1S/C9H7N/c1-2-6-9-8(4-1)5-3-7-10-9/h1-7H

InChI key

SMWDFEZZVXVKRB-UHFFFAOYSA-N

Gene Information

human ... CYP2D6(1565)

Looking for similar products? Visit Product Comparison Guide

General description

Quinoline is a heterocyclic aromatic organic compound with the chemical formula C9H7N.Quinoline derivatives are prevalent in a variety of pharmacologically active synthetic and natural compounds. Quinolines have antiseptic, antipyretic, and antiperiodic properties and are used as antimalarials and for preparing other antimalarial drugs.

Application

Quinoline and its derivatives are widely used as antibiotics, alkaloids, dyes, rubber chemicals, and flavoring agents. Additional industrial applications include their use as corrosion inhibitors and as solvents for resins and terpenes, and in transition-metal complex catalysis for uniform polymerization and luminescence chemistry. They are also used in manufacturing oil soluble dyes, food colorants, pharmaceuticals, pH indicators and other organic compounds. Quinoline is a catabolite of tryptophan, a fundamental structure in some antihypertensive agents such as the peripheral vasodilators prazosin and doxazosin.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yeongkwon Son et al.
International journal of environmental research and public health, 17(8) (2020-04-23)
Dangerous levels of harmful chemicals in electronic cigarette (e-cigarette) aerosols were reported by several studies, but variability in e-cigarette design and use patterns, and a rapid development of new devices, such as JUUL, hamper efforts to develop standardized testing protocols
Nanotube composite carbon fibers
Andrews R, et al.
Applied Physics Letters, 75(9), 1329-1331 (1999)
Marella A, et al.
Saudi Pharmaceutical Journal : SPJ : The Official Publication of the Saudi Pharmaceutical Society, 21(1), 1-2 (2013)
Margarita A Gonzalez et al.
Inorganic chemistry, 51(21), 11930-11940 (2012-10-24)
The unusual role of CO as a signaling molecule in several physiological pathways has spurred research in the area of synthesizing new CO-releasing molecules (CORMs) as exogenous CO donors. Auxiliary control on CO delivery can be achieved if CO can
Yoshiaki Yasutake et al.
FEBS letters, 588(1), 105-110 (2013-11-26)
The cytochrome P450 RauA from Rhodococcus erythropolis JCM 6824 catalyzes the hydroxylation of a nitrogen atom in the quinolone ring of aurachin, thereby conferring strong antibiotic activity on the aurachin alkaloid. Here, we report the crystal structure of RauA in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service