Skip to Content
Merck
All Photos(1)

Key Documents

238082

Sigma-Aldrich

4-Butoxybenzaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OC6H4CHO
CAS Number:
Molecular Weight:
178.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.539 (lit.)

bp

285 °C (lit.)

density

1.031 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

CCCCOc1ccc(C=O)cc1

InChI

1S/C11H14O2/c1-2-3-8-13-11-6-4-10(9-12)5-7-11/h4-7,9H,2-3,8H2,1H3

InChI key

XHWMNHADTZZHGI-UHFFFAOYSA-N

General description

Kinetic constant for the inhibition of the diphenolase activity of mushroom tyrosinase by 4-butoxybenzaldehyde has been evaluated.

Application

4-Butoxybenzaldehyde has been used in the synthesis of:
  • 6-amino-4-(4-butoxyphenyl)-3,5-dicyanopyridine-2(1H)-thione
  • 16-(p-butoxybenzylidene)androsta-1,4-diene-3,17-dione via condensation reaction with androsta-1,4-diene-3,17-dione

Legal Information

Darkens in storage with no loss in purity

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

16-(p-Butoxybenzylidene) androsta-1, 4-diene-3, 17-dione.
Ogawa K, et al.
Acta Crystallographica Section C, Crystal Structure Communications, 48(7), 1359-1361 (1992)
Michael reaction in synthesis of 6-amino-4-(4-butoxyphenyl)-3, 5-dicyanopyridine-2 (1H)-thione.
Dyachenko VD and Litvinov VP.
Chemistry of Heterocyclic Compounds, 34(2), 188-194 (1998)
Dalila Rocco et al.
Molecules (Basel, Switzerland), 24(9) (2019-05-12)
The preparation of 24-functionalized 12,22:26,32-terpyridines (4'-functionalized 3,2:6',3''-terpyridines) by the reaction of three 4-alkoxybenzaldehydes with 3-acetylpyridine and ammonia was investigated; under identical reaction conditions, two (R = nC4H9, C2H5) gave the expected products whereas a third (R = nC3H7) gave only
M Jiménez et al.
Journal of agricultural and food chemistry, 49(8), 4060-4063 (2001-08-22)
A kinetic study of the inhibition of mushroom tyrosinase by 4-substituted benzaldehydes showed that these compounds behave as classical competitive inhibitors, inhibiting the oxidation of L-3,4-dihydroxyphenylalanine (L-DOPA) by mushroom tyrosinase (o-diphenolase activity). The kinetic parameter (K(I)) characterizing this inhibition was
Naoko Ueno et al.
Langmuir : the ACS journal of surfaces and colloids, 33(22), 5393-5397 (2017-05-16)
We evaluated the speed profile of self-propelled underwater oil droplets comprising a hydrophobic aldehyde derivative in terms of their diameter and the surrounding surfactant concentration using a microfluidic device. We found that the speed of the oil droplets is dependent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service