Skip to Content
Merck
All Photos(2)

Key Documents

233552

Sigma-Aldrich

4-Tetradecylaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13C6H4NH2
CAS Number:
Molecular Weight:
289.50
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

46-49 °C (lit.)

SMILES string

CCCCCCCCCCCCCCc1ccc(N)cc1

InChI

1S/C20H35N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-19-15-17-20(21)18-16-19/h15-18H,2-14,21H2,1H3

InChI key

WKYFQPQIOXXPGE-UHFFFAOYSA-N

Application

4-Tetradecylaniline has been used:
  • as standard during the quantitative determination of trans content of fats and oils and margarine by FTIR spectroscopy
  • to investigate the quenching of the triplet state of n-alkyl 3-nitrophenyl ethers in aqueous micellar sodium dodecyl sulfate (SDS) solutions by laser flash photolysis
  • in amidation and esterification of oxidised single-walled carbon nanotubes (SWCNTs)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trans fatty acid determination in fats and margarine by fourier transform infrared spectroscopy using a disposable infrared card.
Ma K, et al.
Journal of the American Oil Chemists' Society, 76(12), 1399-1404 (1999)
Prabhpreet Singh et al.
Chemical Society reviews, 38(8), 2214-2230 (2009-07-23)
Since carbon nanotubes (CNTs) display unique structures and remarkable physical properties, a variety of applications have emerged in both materials and life sciences. In terms of applications, the functionalisation of nanotubes is extremely important, as it increases their solubility and
Photoreactions of n-alkyl-3-nitrophenyl ethers with aromatic amines in SDS micelles: A laser flash photolysis study.
Rigoli IC, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 222(1), 34-39 (2011)
Kristiana T Santoso et al.
Bioorganic & medicinal chemistry, 27(16), 3532-3545 (2019-07-03)
A library of thirty-two quinolinequinones (QQs) with various amine substituents at the 6- and 7-positions were synthesised efficiently and in good yields for evaluation as potential anti-tuberculosis agents. Mycobacterium tuberculosis growth inhibition assays demonstrated that QQs bearing moderate length alkyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service