Skip to Content
Merck
All Photos(1)

Key Documents

222887

Sigma-Aldrich

1-Pentadecene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12CH=CH2
CAS Number:
Molecular Weight:
210.40
Beilstein:
1751011
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.439 (lit.)

bp

268-269 °C (lit.)

mp

−4 °C (lit.)

density

0.775 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCC=C

InChI

1S/C15H30/c1-3-5-7-9-11-13-15-14-12-10-8-6-4-2/h3H,1,4-15H2,2H3

InChI key

PJLHTVIBELQURV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Pentadecene serves as a growth medium for the strain CV2803T. It is a common semiochemical of flour beetles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristiana Cravo-Laureau et al.
Comptes rendus biologies, 327(7), 687-694 (2004-09-04)
The anaerobic degradation of n-alkenes by a sulphate-reducing bacterium Desulfatibacillum aliphaticivorans strain CV2803T was investigated. Results suggest that enzymes required for alkene degradation are inducible. Moreover, total cellular fatty acids of strain CV2803T were predominantly C-odd when the strain was
Ludovic Arnaud et al.
Journal of chemical ecology, 28(3), 523-532 (2002-04-12)
Flour beetles are cosmopolitan and common pests in grain stores and flour mills. Their ability to exploit a wide variety of stored products has contributed to their status as major pests of stored food. Although it was previously reported that
Jaimie Gibbons et al.
AMB Express, 8(1), 167-167 (2018-10-15)
Cyanobacteria photosynthetically produce long-chain hydrocarbons, which are considered as infrastructure-compatible biofuels. However, native cyanobacteria do not produce these hydrocarbons at sufficient rates or yields to warrant commercial deployment. This research sought to identify specific genes required for photosynthetic production of
Vincent Grossi et al.
Applied and environmental microbiology, 81(9), 3157-3168 (2015-03-01)
Bacterial glycerol ether lipids (alkylglycerols) have received increasing attention during the last decades, notably due to their potential role in cell resistance or adaptation to adverse environmental conditions. Major uncertainties remain, however, regarding the origin, biosynthesis, and modes of formation
Geoffrey K Yeh et al.
The journal of physical chemistry. A, 119(43), 10684-10696 (2015-10-06)
The linear C15 alkene, 1-pentadecene, was reacted with NO3 radicals in a Teflon environmental chamber and yields of secondary organic aerosol (SOA) and particulate β-hydroxynitrates, β-carbonylnitrates, and organic peroxides (β-nitrooxyhydroperoxides + dinitrooxyperoxides) were quantified using a variety of methods. Reaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service