Skip to Content
Merck
All Photos(2)

Documents

180076

Sigma-Aldrich

β-Methylphenethylamine

99%

Synonym(s):

beta-Methylphenethylamine, 1-Amino-2-phenylpropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)CH2NH2
CAS Number:
Molecular Weight:
135.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.524 (lit.)

bp

80 °C/10 mmHg (lit.)

density

0.93 g/mL at 25 °C (lit.)

SMILES string

CC(CN)c1ccccc1

InChI

1S/C9H13N/c1-8(7-10)9-5-3-2-4-6-9/h2-6,8H,7,10H2,1H3

InChI key

AXORVIZLPOGIRG-UHFFFAOYSA-N

General description

β-Methylphenethylamine is considered to be a positional isomer of amphetamine.

Application

β-Methylphenethylamine was used for molecular imprinting and to study the space filling models depicting minimized structures for each enantiomer.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rahul S Pawar et al.
Journal of pharmaceutical and biomedical analysis, 88, 457-466 (2013-11-02)
Extracts of Acacia rigidula leaves are used in weight-loss products sold in vitamin shops and over the internet with little or no published data about their potential biological effects. In our chemical investigations on authenticated A. rigidula plant material, we
D A Spivak et al.
The Analyst, 126(6), 793-797 (2001-07-12)
Although molecular imprinting is a widely accepted method for producing template specific polymers, the general rules for prediction and control of the binding and catalytic properties of these materials are still not fully understood. One reason for this is the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service