Skip to Content
Merck
All Photos(1)

Key Documents

167606

Sigma-Aldrich

3-Chloro-2-butanone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CHClCOCH3
CAS Number:
Molecular Weight:
106.55
Beilstein:
385637
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.421 (lit.)

bp

114-117 °C (lit.)

density

1.055 g/mL at 25 °C (lit.)

functional group

chloro
ketone

SMILES string

CC(Cl)C(C)=O

InChI

1S/C4H7ClO/c1-3(5)4(2)6/h3H,1-2H3

InChI key

OIMRLHCSLQUXLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Chloro-2-butanone reacts with 1,4-dianion of acetophenone N-ethoxycarbonylhydrazone to yield pyrazoline derivatives.

Application

3-Chloro-2-butanone was used in production of chiral alcohols from acetophenone derivatives, β-ketoesters and N-Boc-3-pyrrolidinone by recombinant E. coli cells. It was used in the synthesis of carbene precursor, 3-aryl-4,5-dimethylthiazolium chloride.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Georgios C Vougioukalakis et al.
Journal of the American Chemical Society, 130(7), 2234-2245 (2008-01-29)
A new family of ruthenium-based olefin metathesis catalysts bearing a series of thiazole-2-ylidene ligands has been prepared. These complexes are readily accessible in one step from commercially available (PCy3)2Cl2Ru=CHPh or (PCy3)Cl2Ru=CH(o-iPrO-Ph) and have been fully characterized. The X-ray crystal structures
Reaction of α-chloroketones with 1, 4-dianion of acetophenone n-ethoxy-carbonylhydrazone.
Matsumura N, et al.
Tetrahedron Letters, 25(40), 4529-4532 (1984)
Nobuya Itoh et al.
European journal of biochemistry, 269(9), 2394-2402 (2002-05-03)
Phenylacetaldehyde reductase (PAR) produced by styrene-assimilating Corynebacterium strain ST-10 was used to synthesize chiral alcohols. This enzyme with a broad substrate range reduced various prochiral aromatic ketones and beta-ketoesters to yield optically active secondary alcohols with an enantiomeric purity of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service