Skip to Content
Merck
All Photos(1)

Key Documents

159387

Sigma-Aldrich

3-(Trifluoromethyl)phenyl isocyanate

97%

Synonym(s):

α,α,α-Trifluoro-m-tolyl isocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4NCO
CAS Number:
Molecular Weight:
187.12
Beilstein:
744880
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

2.44 psi ( 20 °C)

Assay

97%

refractive index

n20/D 1.467 (lit.)

bp

54 °C/11 mmHg (lit.)

density

1.359 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cccc(c1)N=C=O

InChI

1S/C8H4F3NO/c9-8(10,11)6-2-1-3-7(4-6)12-5-13/h1-4H

InChI key

SXJYSIBLFGQAND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-(Trifluoromethyl)phenyl isocyanate (α,α,α-Trifluoro-m-tolyl isocyanate) was used in the preparation of (S)-3-hydroxymethyl-2-methoxymethoxy-2′-(3-(3-(trifluoromethyl)phenyl)uryl-benzyl)-1,1′-binaphthalene. It was also used in the preparation of cholesteryl trifluoromethylphenyl carbamate.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service