Skip to Content
Merck
All Photos(1)

Key Documents

15450

Sigma-Aldrich

N,N′-Dimethylurea

(sym.), ≥99% (from N)

Synonym(s):

DMU, 1,3-Dimethylurea

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3NH)2CO
CAS Number:
Molecular Weight:
88.11
Beilstein:
1740672
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99% (from N)

form

solid

reaction suitability

reagent type: ligand
reaction type: C-H Activation

bp

268-270 °C (lit.)

mp

101-104 °C (lit.)
103-107 °C

SMILES string

CNC(=O)NC

InChI

1S/C3H8N2O/c1-4-3(6)5-2/h1-2H3,(H2,4,5,6)

InChI key

MGJKQDOBUOMPEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N′-Dimethylurea can be used:
  • As a starting material to synthesize N,N′-dimethyl-6-amino uracil.
  • In combination with β-cyclodextrin derivatives, to form low melting mixtures (LMMs), which can be used as solvents for hydroformylation and Tsuji-Trost reactions.
  • To synthesize N,N′-disubstituted-4-aryl-3,4-dihydropyrimidinones via Biginelli condensation under solvent-free conditions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactions of 1, 3-Dimethyl-5, 6-diaminouracil.
Blicke F F and Godt Jr H C
Journal of the American Chemical Society, 76(10), 2798-2800 (1954)
Hyun-Woong Cho et al.
PloS one, 14(6), e0217745-e0217745 (2019-06-21)
The aim of this study was to investigate the short-term efficacy and safety of Poly-gamma-glutamic acid (γ-PGA) and the immunologic changes in patients with CIN 1. Participants were randomly assigned to one of two groups and orally treated with placebo
Low melting mixtures based on β-cyclodextrin derivatives and N, N′-dimethylurea as solvents for sustainable catalytic processes.
Jerome F, et al.
Green Chemistry, 16(8), 3876-3880 (2014)
Dowex-promoted general synthesis of N, N'-disubstituted-4-aryl-3, 4-dihydropyrimidinones using a solvent-free Biginelli condensation protocol.
Singh K, et al.
Tetrahedron Letters, 47(25), 4205-4207 (2006)
Dimethylurea: a radical scavenger that protects isolated pancreatic islets from the effects of alloxan and dihydroxyfumarate exposure.
L J Fischer et al.
Life sciences, 26(17), 1405-1409 (1980-04-28)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service