All Photos(3)
About This Item
Linear Formula:
BrC6H4NHNH2·HCl
CAS Number:
Molecular Weight:
223.50
Beilstein:
3628612
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
mp
189 °C (dec.) (lit.)
functional group
bromo
hydrazine
SMILES string
Cl.NNc1ccccc1Br
InChI
1S/C6H7BrN2.ClH/c7-5-3-1-2-4-6(5)9-8;/h1-4,9H,8H2;1H
InChI key
PHCYUJRYSFMJMG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
2-Bromophenylhydrazine hydrochloride was used to prepare 6-(3-bromophenylhydrazono)-7,8,9,11-tetrahydropyrido[2,1-b]quinazoline-11-one.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis and COX inhibitory activities of rutaecarpine derivatives.
Bull. Korean Chem. Soc., 26(12), 1975-1975 (2005)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service