Merck
  • Fullerene-C60 derivatives prevent UV-irradiation/ TiO2-induced cytotoxicity on keratinocytes and 3D-skin tissues through antioxidant actions.
Fullerene-C60 derivatives prevent UV-irradiation/ TiO2-induced cytotoxicity on keratinocytes and 3D-skin tissues through antioxidant actions.
Journal of nanoscience and nanotechnology (2014-04-17)
Shinya Kato, Hisae Aoshima, Yasukazu Saitoh, Nobuhiko Miwa
ABSTRACT
Microcorpuscular titanium dioxide (TiO2), a useful sunscreen agent, photocatalyzes generation of reactive oxygen species (ROS). We assessed protective effects of fullerene-C60 derivatives or microcolloidal platinum (Pt) against ultraviolet ray (UV)-irradiation in the presence of TiO2 in vitro. UV-irradiation (8 J/cm2, mixed UVA and UVB) in the presence of 15 ppm TiO2 on HaCaT keratinocytes decreased cell viability as quantified by WST-1 assay, and increased both intracellular ROS and cell-membrane-lipid peroxidation, as quantified by nitroblue-tetrazolium (NBT) assay and diphenyl-1-pyrenylphosphine (DPPP) assay, respectively, whereas all of three phototoxicity-related symptoms were appreciably repressed almost to UV-unirradiational levels by pretreatment with polyvinylpyrrolidone-entrapped fullerene-C60 (C60/PVP) or fullerene-C60 dissolved in squalane (C60/Sqn) in a dose-dependent manner of C60, but scarcely by PVP alone or Sqn alone. In contrast, Pt repressed intracellular ROS generation, but did not prevent either peroxidation of cell-membrane-lipid or cell mortality. Then in the epidermis of 3-dimensional human skin tissue model, UV-irradiation in the presence of TiO2 extensively induced two symptoms such as ROS-generation around perinuclear regions and membrane-lipid peroxidation, both of which were repressed by C60/PVP or C60/Sqn, whereas Pt did not prevent membrane-lipid peroxidation adequately. Thus the advantageous application of the lipophilic antioxidant fullerene-C60 which effectively protects cell membrane against peroxidation. In conclusion, fullerene-C60 can be expected to serve as an antioxidant for scavenging of TiO2-photocatalyzed ROS in the skin surface, and therefore provide a functional improvement of TiO2-containing sunscreens.
Sigma-Aldrich
Fullerene-C60, 99.5%
Sigma-Aldrich
Fullerene-C60, sublimed, 99.9%
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5%
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~100 nm × 10 μm
Sigma-Aldrich
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~10 nm × 10 μm
Sigma-Aldrich
Fullerene-C60, 98%
Sigma-Aldrich
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, sponge, 1-20 mm, 99.5% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
Titanium, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.995% trace metals basis
Titanium, IRMM®, certified reference material, 0.1 mm foil
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, ≥99.99% trace metals basis
Titanium, rod, 100mm, diameter 20mm, annealed, 99.6+%
Sigma-Aldrich
Titanium(IV) oxide, ReagentPlus®, ≥99%
Titanium, tube, 100mm, outside diameter 12.7mm, inside diameter 10.9mm, wall thickness 0.9mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 6.35mm, inside diameter 4.57mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, rod, 50mm, diameter 16mm, as drawn, 99.99+%
Titanium, rod, 500mm, diameter 2mm, as drawn, 99.99+%
Titanium, tube, 200mm, outside diameter 9.5mm, inside diameter 8.2mm, wall thickness 0.65mm, annealed, 99.6+%
Titanium, rod, 50mm, diameter 8.0mm, as drawn, 99.99+%