Merck
  • BNIP3 promotes HIF-1α-driven melanoma growth by curbing intracellular iron homeostasis.
BNIP3 promotes HIF-1α-driven melanoma growth by curbing intracellular iron homeostasis.
The EMBO journal (2021-05-02)
Mónica Vara-Pérez, Matteo Rossi, Chris Van den Haute, Hannelore Maes, Maria Livia Sassano, Vivek Venkataramani, Bernhard Michalke, Erminia Romano, Kristine Rillaerts, Abhishek D Garg, Corentin Schepkens, Francesca M Bosisio, Jasper Wouters, Ana Isabel Oliveira, Peter Vangheluwe, Wim Annaert, Johannes V Swinnen, Jean Marie Colet, Joost J van den Oord, Sarah-Maria Fendt, Massimiliano Mazzone, Patrizia Agostinis
ABSTRACT
BNIP3 is a mitophagy receptor with context-dependent roles in cancer, but whether and how it modulates melanoma growth in vivo remains unknown. Here, we found that elevated BNIP3 levels correlated with poorer melanoma patient's survival and depletion of BNIP3 in B16-F10 melanoma cells compromised tumor growth in vivo. BNIP3 depletion halted mitophagy and enforced a PHD2-mediated downregulation of HIF-1α and its glycolytic program both in vitro and in vivo. Mechanistically, we found that BNIP3-deprived melanoma cells displayed increased intracellular iron levels caused by heightened NCOA4-mediated ferritinophagy, which fostered PHD2-mediated HIF-1α destabilization. These effects were not phenocopied by ATG5 or NIX silencing. Restoring HIF-1α levels in BNIP3-depleted melanoma cells rescued their metabolic phenotype and tumor growth in vivo, but did not affect NCOA4 turnover, underscoring that these BNIP3 effects are not secondary to HIF-1α. These results unravel an unexpected role of BNIP3 as upstream regulator of the pro-tumorigenic HIF-1α glycolytic program in melanoma cells.
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-BNIP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid