Skip to Content
Merck
All Photos(1)

Key Documents

1091028

USP

Capric alcohol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1-Decanol, n-Decyl alcohol, Alcohol C10

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9OH
CAS Number:
Molecular Weight:
158.28
Beilstein:
1735221
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

4.5 (vs air)

vapor pressure

1 mmHg ( 70 °C)
8.25 mmHg ( 100 °C)

API family

capric alcohol

autoignition temp.

550 °F

manufacturer/tradename

USP

refractive index

n20/D 1.437 (lit.)

bp

231 °C (lit.)

mp

5-7 °C (lit.)

density

0.829 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCCCO

InChI

1S/C10H22O/c1-2-3-4-5-6-7-8-9-10-11/h11H,2-10H2,1H3

InChI key

MWKFXSUHUHTGQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Capric alcohol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Cocoyl Caprylocaprate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

203.0 °F - Pensky-Martens closed cup

Flash Point(C)

95 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service