Skip to Content
Merck
All Photos(1)

Key Documents

PHR9050

Supelco

4-[2-(tert-Butylamino)-1-methoxyethyl]-2-(hydroxymethyl)phenol

reference material, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Salbutamol Impurity A (PhEur), 5-[(1RS)-2-[(1,1-Dimethylethyl)amino]-1-methoxyethyl]-2-hydroxyphenyl]methanol, 5-[2-[(1,1-Dimethylethyl)amino]-1-methoxyethyl]-2-hydroxybenzenemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H23NO3
CAS Number:
Molecular Weight:
253.34
Beilstein:
14479390
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

reference material

Quality Level

API family

salbutamol

Assay

≥97.0% (HPLC)

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

application(s)

pharmaceutical small molecule

SMILES string

OCC1=C(O)C=CC(C(OC)CNC(C)(C)C)=C1

InChI

1S/C14H23NO3/c1-14(2,3)15-8-13(18-4)10-5-6-12(17)11(7-10)9-16/h5-7,13,15-17H,8-9H2,1-4H3

InChI key

UMHASVFLCHGDPW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Application

These Reference Materials are suitable for use in several analytical applications including, but not limited to, method development for qualitative and quantitative analyses, daily calibration, and routine quality control testing.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service