Skip to Content
Merck
All Photos(8)

Key Documents

ZRB2001

Sigma-Aldrich

Anti-Nur77/NR4A1 Antibody, clone 1F2 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Early response protein NAK1, Nuclear hormone receptor NUR/77, Nuclear receptor subfamily 4 group A member 1, Nur77, Orphan nuclear receptor HMR, Orphan nuclear receptor TR3, ST-59, Testicular receptor 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
conjugate:
unconjugated
application:
ELISA
IHC
WB
affinity binding assay
clone:
1F2, recombinant monoclonal
species reactivity:
human
citations:
technique(s):
ELISA: suitable
affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1F2, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 64.46 kDa
observed mol wt ~60 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1F2 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Nur77/NR4A1. It targets an epitope within 17 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the N-terminal half of human Nur77/NR4A1.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected Nur77/NR4A1 in HeLa cell lysate.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Nur77/NR4A1 in Human liver tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound Nur77/NR4A1 peptide with a KD of 2.4 x 10-6 in an affinity binding assay.

Enzyme Immunoassay Analysis (ELISA): A serial of dilutions from a representative lot detected recombinant Human Nur77/NR4A1.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

NuCLEAR receptor subfamily 4 group A member 1 (UniProt: P22736; also known as Early response protein NAK1, NuCLEAR hormone receptor NUR/77, Nur77, Orphan NuCLEAR receptor HMR, Orphan NuCLEAR receptor TR3, ST-59, Testicular receptor 3) is encoded by the NR4A1 (also known as GFRP1, HMR, NAK1) gene (Gene ID: 3164) in human. Nur77 is an orphan NuCLEAR receptor that may ACT® concomitantly with NURR1 in regulating the expression of delayed-early genes during liver regeneration. Its expression is observed in fetal muscle and in adult liver, brain, and thyroid and is reported to be induced by growth stimulating agents. It binds the NGFI-B response Element (NBRE) 5′-AAAAGGTCA-3′. Its DNA binding region is localized to amino acids 264-339. It contains two zinc finger domains (aa 267-287 and 303-327). Nur77 is shown to heterodimerize with retinoid X receptor alpha and is actively transported from the nucleus to the mitochondria in cells undergoing apoptosis. It binds to Bcl-2 and triggers a conformational change that exposes the pro-apoptotic BH3 domain. This converts Bcl-2 from an anti-apoptotic to a pro-apoptotic factor. In several cancers the NuCLEAR-mitochondrial translocation of Nur77/TR3 is defective, which prevents apoptosis. Three isoforms of Nur77 have been described that are produced by alternative splicing. It is acetylated by p300/CBP that increases its stability. It can also undergo phosphorylation at Serine 351 by RPS6KA1 and RPS6KA3 in response to mitogenic or stress stimuli.  This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Liu, J., et al. (2017). Oncotarget. 8(41); 69731-69745; Lin, B., et al. (2004). Cell. 116(4); 527-540).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ACT is a registered trademark of Merck KGaA, Darmstadt, Germany
Affinity is a trademark of Mine Safety Appliances Co.
Element is a trademark of Corning, Inc.
NuCLEAR is a trademark of Sigma-Aldrich Co. LLC
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service