Z763373
Orbi-Shaker™ XL orbital shaker with flat mat platform
AC/DC input 115 V AC
Synonym(s):
orbital mixers, orbital shakers, stirring plates
About This Item
Recommended Products
manufacturer/tradename
Benchmark Scientific BT1011
parameter
30-300 rpm speed
overall L × W × H
18.5 in. (47 cm) × 21.25 in. (54 cm) × 5 in. (12.5 cm)
platform L × W
17.8 in. (45 cm) × 17.8 in. (45 cm)
weight
30 kg
application(s)
PFAS testing
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service