Skip to Content
Merck
All Photos(1)

Key Documents

Z640417

Eppendorf® epT.I.P.S. Reloads

volume range 2-200 μL

Synonym(s):

Eppendorf reloadable pipette tips, pipetting tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.01
L:
53 mm
maximum volume:
200 μL
volume range:
2-200 μL
sterility:
non-sterile
feature:
barrier: no
packaging:
pack of 960 (10 trays of 96 tips)
manufacturer/tradename:
Eppendorf® 0030 073.428
Eppendorf® 022491539

material

yellow tip

sterility

non-sterile

feature

barrier: no

packaging

pack of 960 (10 trays of 96 tips)

manufacturer/tradename

Eppendorf® 0030 073.428
Eppendorf® 022491539

L

53 mm

maximum volume

200 μL

volume range

2-200 μL

Looking for similar products? Visit Product Comparison Guide

General description

Part of the ep T.I.P.S. box and reload system for contamination-free handling of refill trays, with plastic lids protecting the trays from contact during transfer to the working box.
Trays of tips which may be autoclaved in their packaging and stored clean for later use. Tips may be used straight from the reload tray, or loaded into ep T.I.P.S box (purchase separately)
Reloads are trays specifically designed to contain pipette tips for refilling a reusable box. The refill trays, which also house the pipette tips, can undergo autoclaving within the box at temperatures of up to 121 °C, ensuring reliable sterilization. Depending on the pipette tip volume, the refill trays can either be tightly packed together or stacked, providing efficient storage and organizational options. It is vital to remember that prior to using the pipette tips, both the refill trays and the tips themselves must be correctly inserted into the reusable box, guaranteeing ease of access and convenience during pipetting tasks.

Application

Eppendorf® epT.I.P.S. Reloads are suitable:
  • for general laboratory applications, including low-contamination transfer of liquids
  • for gentle mixing of cell suspension
  • for resuspending the cells from the colony in the well of the selection plate

Features and Benefits

  • Reduced waste in comparison to disposable racks
  • Packaging for the refill system varies based on tip size, including dual-sided or stacked options
  • Color-coded trays enable easy identification of volume and pipettes
  • Single-use pipette tips with a universal cone for compatibility
  • User-friendly and easy to handle
  • Offered in various purity grades
  • Volume marking allows for convenient visual inspection
  • Simple attachment and ejection with minimal force required
  • Complete sealing ensures reliable performance
  • Provides maximum precision and accuracy
  • Long pipette tips are available for specific volume sizes

Other Notes

The epT.I.P.S. pipette tips are intended for single-use only and should be used exclusively by qualified personnel.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detection of Loss of Heterozygosity in Tk-Deficient Mutants from L5178Y Tk+/--3.7.2C Mouse Lymphoma Cells
Guo X, et al.
Methods in Molecular Biology (2020)
Measuring key human carbohydrate digestive enzyme activities using high-performance anion-exchange chromatography with pulsed amperometric detection
Barber E, et al.
Nature Protocols (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service