WPB01341WA
Whirl-Pak® Flat Wire, Write-On Bag
532 mL capacity(18 oz)
Synonym(s):
sample bag with label, sampling bag
All Photos(1)
About This Item
UNSPSC Code:
24111500
NACRES:
NB.35
Recommended Products
packaging
pkg of 500 ea
manufacturer/tradename
B01341WA (Nasco)
W × L × thickness
11.5 cm × 23 cm × 0.064 mm
capacity
532 mL × 18 oz
532 mL capacity(18 oz)
Related Categories
General description
These bags are made with one flat wire, rather than the traditional round wires. The flat wire provides a thicker tab, resulting in a heavier closure which is preferred in some applications. The aluminum wire features “PUNCTURE PROOF TABS,” so the wire ends are completely covered, eliminating any sharp, cutting, exposed edges. The aluminum will not rust when coming in contact with water or other liquids.
The white, write-on strip makes sample identification easier and more convenient. It′s easy to write on this white strip using an ordinary ballpoint pen, pencil, or felt-tip marker. The paper-like finish virtually eliminates ink smearing, and provides a surface that will accept a variety of writing instruments.
The white, write-on strip makes sample identification easier and more convenient. It′s easy to write on this white strip using an ordinary ballpoint pen, pencil, or felt-tip marker. The paper-like finish virtually eliminates ink smearing, and provides a surface that will accept a variety of writing instruments.
Legal Information
Whirl-pak is a registered trademark of The Aristotle Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)