Skip to Content
Merck
All Photos(1)

Key Documents

SML3331

Sigma-Aldrich

Triheptanoin

≥95% (HPLC)

Synonym(s):

Dermofeel TC 7, Glycerol trienanthate, Glyceryl triheptanoate, Heptanoic acid, 1,1′,1′′-(1,2,3-propanetriyl) ester, Heptanoic acid, 1,2,3-propanetriyl ester (9CI), Heptanoin, tri- (6CI,7CI,8CI), Lanol 37T, Trienanthoin, Triheptanoic glyceride, Trioenanthoin, UX007

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H44O6
CAS Number:
Molecular Weight:
428.60
MDL number:
UNSPSC Code:
23201100
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

oil

color

colorless to brown

storage temp.

−20°C

SMILES string

O(C(COC(=O)CCCCCC)COC(=O)CCCCCC)C(=O)CCCCCC

InChI

1S/C24H44O6/c1-4-7-10-13-16-22(25)28-19-21(30-24(27)18-15-12-9-6-3)20-29-23(26)17-14-11-8-5-2/h21H,4-20H2,1-3H3

InChI key

PJHKBYALYHRYSK-UHFFFAOYSA-N

Biochem/physiol Actions

Orally available triglyceride that provides 3 molar eq of saturated heptanoic acid in vivo to replenish reduced tricarboxylic acid (TCA; Krebs)-cycle intermediates.
Triheptanoin is a triglyceride with 3 molar eq. of saturated heptanoic acid (C7:0). The even-chain FA does not occur naturally and is able to bypass metabolic block induced by enzyme deficiency and to replenish reduced tricarboxylic acid/TCA-cycle (Krebs cycle or citric acid cycle/CAC) intermediates (anaplerosis) in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

404.6 °F - open cup

Flash Point(C)

207 °C - open cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Tucci et al.
Journal of lipid research, 58(1), 196-207 (2016-11-26)
A rather new approach in the treatment of long-chain fatty acid oxidation disorders is represented by triheptanoin, a triglyceride with three medium-odd-chain heptanoic acids (C7), due to its anaplerotic potential. We here investigate the effects of a 1-year triheptanoin-based diet
T M Schwarzkopf et al.
Neuroscience, 300, 201-209 (2015-05-20)
Triheptanoin, an oily substance, consists of glycerol bound to three molecules of heptanoic acid, a C7 odd-chain fatty acid. A triheptanoin-rich diet has anaplerotic effects because heptanoate metabolism yields succinate which delivers substrates to the Krebs cycle. While previous studies
Ingrid Sofia Vieira de Melo et al.
Nutricion hospitalaria, 32(1), 265-269 (2015-08-12)
the aim of this study was to evaluate the influence of consumption of a ketogenic diet supplemented with triheptanoin, a medium-chain anaplerotic triacylglycerol, on the liver fatty acid profile of Wistar rats. three groups of male Wistar rats (n =
Keri J Fogle et al.
Molecular genetics and metabolism, 126(4), 439-447 (2019-01-27)
Seizures are a feature not only of the many forms of epilepsy, but also of global metabolic diseases such as mitochondrial encephalomyopathy (ME) and glycolytic enzymopathy (GE). Modern anti-epileptic drugs (AEDs) are successful in many cases, but some patients are
Tesfaye W Tefera et al.
PloS one, 11(8), e0161816-e0161816 (2016-08-27)
There is increasing evidence that energy metabolism is disturbed in Amyotrophic Lateral Sclerosis (ALS) patients and animal models. Treatment with triheptanoin, the triglyceride of heptanoate, is a promising approach to provide alternative fuel to improve oxidative phosphorylation and aid ATP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service