Skip to Content
Merck
All Photos(1)

Key Documents

SML3278

Sigma-Aldrich

AC-262536

≥95% (HPLC)

Synonym(s):

154BG31, 4-(3-endo-Hydroxy-8-azabicyclo[3.2.1]oct-8-yl)naphthalene-1-carbonitrile, 4-[(3-endo)-3-Hydroxy-8-azabicyclo[3.2.1]oct-8-yl]-1-naphthalenecarbonitrile, AC 262536, AC262536

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N2O
CAS Number:
Molecular Weight:
278.35
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

Biochem/physiol Actions

AC-262536 is a non-steroidal, tissue-selective androgen receptor (AR) modulator (SARM) that targets AR with high-affinity (pKi = 8.3 vs. 8.9/testosterone), potency (NIH-3T3/MDA-kb2 reporter assay pEC50 = 7.9/8.8 vs. 8.7/8.5/testosterone) and partial agonist activity (Emax = 67% and 72% of testosterone Emax, respectively by NIH-3T3 or MDA-kb2 assay) and high selectivity (no significant affinity for 47 other human nuclear receptors). AC-262536 antagonizes 10 nM DHT-induced LNCaP proliferation (by 47.2% at 100 nM) and prevents castration-induced LH upregulation in rats in vivo (EC50 = 2.8 mg/kg) with greatly reduced adverse effects than testosterone propionate (TP).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrice Piu et al.
The Journal of steroid biochemistry and molecular biology, 109(1-2), 129-137 (2008-01-01)
Because of the limitations and liabilities of current testosterone therapies, non-steroidal tissue-selective androgen receptor modulators may provide a clinically meaningful advance in therapy. Using a functional cell-based assay AC-262536 was identified as a potent and selective AR ligand, with partial
Tim Sobolevsky et al.
Drug testing and analysis, 13(2), 283-298 (2020-08-28)
In the recent years, a lot of effort was put into the development of multiclass initial testing procedures (ITP) to streamline analytical workflow in antidoping laboratories. Here, a high-throughput assay based on liquid chromatography-triple quadrupole mass spectrometry suitable for use
Charlotte Cutler et al.
Drug testing and analysis, 13(2), 369-385 (2020-09-23)
AC-262536 is one of a number of selective androgen receptor modulators that are being developed by the pharmaceutical industry for treatment of a range of clinical conditions including androgen replacement therapy. Though not available therapeutically, selective androgen receptor modulators are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service