Skip to Content
Merck
All Photos(1)

Key Documents

SML2796

Sigma-Aldrich

PVHD121

≥98% (HPLC)

Synonym(s):

α-(4-Methoxyphenyl)-α-methyl-4-quinazolinemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16N2O2
CAS Number:
Molecular Weight:
280.32
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(C)(C1=NC=NC2=CC=CC=C21)C3=CC=C(C=C3)OC

InChI key

WGDUWVWCDNCAOT-UHFFFAOYSA-N

Biochem/physiol Actions

PVHD121 is potent antimitotic agent that binds to the colchicine site of tubulin. PVHD121 selectively inhibits microtubule formation at centrosomes during mitosis. It delays entry into mitosis causing long lasting mitotic arrest by activation of spindle checkpoint.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenta Kuroiwa et al.
ACS medicinal chemistry letters, 6(3), 287-291 (2015-03-31)
A quinazoline derivative PVHD121 (1a) was shown to have strong antiproliferative activity against various tumor-derived cell lines, including A549 (lung), NCI-H460 (lung), HCT116 (colon), MCF7 (breast), PC3 (prostate), and HeLa (cervical) cells with IC50 values from 0.1 to 0.3 μM.
Jun-Ichi Sawada et al.
Molecular pharmacology, 96(5), 609-618 (2019-09-01)
In the research field of tubulin-binding agents for the development of anticancer agents, hidden targets are emerging as a problem in understanding the exact mechanisms of actions. The quinazoline derivative 1-(4-methoxyphenyl)-1-(quinazolin-4-yl)ethan-1-ol (PVHD121) has anti-cell proliferative activity and inhibits tubulin polymerization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service