Skip to Content
Merck
All Photos(1)

Key Documents

SML2662

Sigma-Aldrich

SPA70

≥98% (HPLC)

Synonym(s):

1-(2,5-Dimethoxyphenyl)-4-[[4-(1,1-dimethylethyl)phenyl]sulfonyl]-5-methyl-1H-12,3-triazole, LC-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25N3O4S
CAS Number:
Molecular Weight:
415.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(c2nn[n](c2C)c3c(ccc(c3)OC)OC)c1ccc(cc1)C(C)(C)C

InChI key

OIOPHIFJQXHPAF-UHFFFAOYSA-N

Biochem/physiol Actions

SPA70 is a cell penetrant, potent and highly selective human pregnane X receptor (hPXR) antagonist that inhibits hPXR in humanized mouse models. It inhibits hPXR-mediated drug metabolism in in primary human hepatocytes and in mouse models.
cell penetrant, potent and highly selective human pregnane X receptor (hPXR) antagonist that inhibits hPXR in humanized mouse models.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergio C Chai et al.
Drug discovery today, 24(3), 906-915 (2019-02-08)
The pregnane X receptor (PXR) and the constitutive androstane receptor (CAR) are ligand-activated nuclear receptors (NRs) that are notorious for their role in drug metabolism, causing unintended drug-drug interactions and decreasing drug efficacy. They control the xenobiotic detoxification system by
Combination of Paclitaxel and PXR Antagonist SPA70 Reverses Paclitaxel-Resistant Non-Small Cell Lung Cancer.
Niu, et al.
Cells, 11 (2022)
Wenwei Lin et al.
Nature communications, 8(1), 741-741 (2017-10-01)
Many drugs bind to and activate human pregnane X receptor (hPXR) to upregulate drug-metabolizing enzymes, resulting in decreased drug efficacy and increased resistance. This suggests that hPXR antagonists have therapeutic value. Here we report that SPA70 is a potent and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service