Skip to Content
Merck
All Photos(1)

Key Documents

SML1854

Sigma-Aldrich

Banoxantrone dihydrochloride

≥98% (HPLC)

Synonym(s):

1,4-Bis[[2-(dimethyloxidoamino)ethyl]amino]-5,8-dihydroxy-9,10-anthracenedione, AQ4N dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28N4O6 · 2HCl
CAS Number:
Molecular Weight:
517.40
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

blue

solubility

H2O: 3 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C1C2=C(C(NCC[N+](C)([O-])C)=CC=C2NCC[N+]([O-])(C)C)C(C3=C(O)C=CC(O)=C31)=O.[H]Cl.[H]Cl

InChI

1S/C22H28N4O6.2ClH/c1-25(2,31)11-9-23-13-5-6-14(24-10-12-26(3,4)32)18-17(13)21(29)19-15(27)7-8-16(28)20(19)22(18)30;;/h5-8,23-24,27-28H,9-12H2,1-4H3;2*1H

InChI key

SBWCPHUXRZRTDP-UHFFFAOYSA-N

Application

Banoxantrone dihydrochloride has been used as an organic ligand during a self-assembled metal-organic coordinated nanoparticle (Cu–OCNP/Lap) synthesis. It has also been used for the preparation of supramolecularly functionalized graphene oxide for hypoxia-activated chemotherapy of cancer.

Biochem/physiol Actions

Banoxantrone (AQ4N) is a hypoxia-activated prodrug of topoisomerase II inhibitor AQ4 (Bioreductive AQ4 precursor).
Banoxantrone dihydrochloride enhances the anti-tumor effect caused by radiation.
Hypoxia-activated prodrug of topoisomerase II inhibitor AQ4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O P Friery et al.
British journal of cancer, 82(8), 1469-1473 (2000-04-26)
The ability of the bioreductive drugs AQ4N and tirapazamine to enhance the anti-tumour effect of cyclophosphamide was assessed in three murine tumour models. In male BDF mice implanted with the T50/80 mammary carcinoma, AQ4N (50-150 mg kg(-1)) in combination with
Olivier Trédan et al.
Cancer research, 69(3), 940-947 (2009-01-30)
Hypoxic tumor cells are likely to be resistant to conventional chemotherapy, in large part because many anticancer drugs are unable to penetrate into poorly oxygenated tumor tissue. Here, we used quantitative immunofluorescence to study the distribution of mitoxantrone and AQ4N
Alshad S Lalani et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 13(7), 2216-2225 (2007-04-04)
The antitumor activities and pharmacokinetics of the hypoxia-activated cytotoxin AQ4N and its metabolites were assessed in several preclinical models of pancreatic cancers. The cytotoxic effects of AQ4N prodrug and its bioreduced form, AQ4, were tested against multiple human tumor cell
Yuan-Fu Ding et al.
Biomaterials science, 9(10), 3804-3813 (2021-04-22)
Nano-graphene oxide (NGO) has attracted increasing attention as an advanced drug delivery system. However, the current surface functionalization and drug-loading of NGO either rely on π-π stacking that is limited to aromatic molecules, or covalent conjugation that requires tedious synthesis.
Yuling He et al.
Biomaterials, 275, 120962-120962 (2021-06-22)
Chemodynamic therapy (CDT) is an ideal therapeutic modality with endogenous H2O2 as stimulus. Most intracellular H2O2 supplement strategies for improving CDT efficiency are strongly rely on oxygen participation, and the hypoxia tumor microenvironment impairs their performance. Here we develop a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service