Skip to Content
Merck
All Photos(1)

Key Documents

SML1353

Sigma-Aldrich

ML351

≥98% (HPLC)

Synonym(s):

5-(Methylamino)-2-(1-naphthalenyl)-4-oxazolecarbonitrile, CID 664510, ML 351, SID 104223766

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11N3O
CAS Number:
Molecular Weight:
249.27
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CNC1=C(C#N)N=C(O1)C2=C3C(C=CC=C3)=CC=C2

InChI

1S/C15H11N3O/c1-17-15-13(9-16)18-14(19-15)12-8-4-6-10-5-2-3-7-11(10)12/h2-8,17H,1H3

InChI key

DYXYXTDIFMDJIR-UHFFFAOYSA-N

Biochem/physiol Actions

ML351 is a cell penetrant, selective and highly potent human lipoxygenase-12/15 (15-Lipoxygenase-1, 12/15-LOX) inhibitor that exhibits protective effects against oxidative glutamate toxicity in mouse neuronal HT22 cells. ML351 reduces infarct size in a mouse model of ischemic stroke.
ML351 is a cell penetrant, selective and highly potent human lipoxygenase-12/15 (15-Lipoxygenase-1, 12/15-LOX) inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sally E Wenzel et al.
Cell, 171(3), 628-641 (2017-10-21)
Ferroptosis is a form of programmed cell death that is pathogenic to several acute and chronic diseases and executed via oxygenation of polyunsaturated phosphatidylethanolamines (PE) by 15-lipoxygenases (15-LO) that normally use free polyunsaturated fatty acids as substrates. Mechanisms of the
Lisa Mayr et al.
Nature communications, 11(1), 1775-1775 (2020-04-15)
The increased incidence of inflammatory bowel disease (IBD) has become a global phenomenon that could be related to adoption of a Western life-style. Westernization of dietary habits is partly characterized by enrichment with the ω-6 polyunsaturated fatty acid (PUFA) arachidonic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service