Skip to Content
Merck
All Photos(7)

Key Documents

SAB4504099

Sigma-Aldrich

Anti-phospho-PKC-pan (pThr497) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-PKCL, Anti-PRKCL, Anti-nPKC-eta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 76 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr497)

General description

Protein kinase C is a member of the cAMP-dependent, cGMP-dependent, and protein kinase C (AGC) superfamily of protein kinases. It has more than14 isoforms. A catalytic domain with an ATP binding site and a substrate-binding site is found in all PKC isoforms.

Immunogen

The antiserum was produced against synthesized peptide derived from human PKC-pan around the phosphorylation site of Thr497.

Immunogen Range: 623-672

Application

Anti-phospho-PKC-pan (pThr497) antibody produced in rabbit has been used in western blot analysis (1:500).

Biochem/physiol Actions

Protein kinase Cs (PKCs) are involved in the growth and function of the gastrointestinal tract and the immune response. PKCs regulates cyclins, cyclin-dependent kinases (cdks), cdk inhibitors, and cell division cycle 25 (cdc25) phosphatases.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genevieve L Wojcik et al.
mBio, 11(1) (2020-02-06)
Diarrhea is a major cause of both morbidity and mortality worldwide, especially among young children. Cryptosporidiosis is a leading cause of diarrhea in children, particularly in South Asia and sub-Saharan Africa, where it is responsible for over 200,000 deaths per
Adrian R Black et al.
Frontiers in immunology, 3, 423-423 (2013-01-22)
A link between T cell proliferation and the protein kinase C (PKC) family of serine/threonine kinases has been recognized for about 30 years. However, despite the wealth of information on PKC-mediated control of, T cell activation, understanding of the effects
Serine and threonine phosphorylation
Plattner F and Bibb JA
Basic Neurochemistry, 467-492 (2012)
Shujie Ning et al.
International journal of molecular sciences, 20(7) (2019-04-17)
Previous studies have demonstrated that monochromatic light affects plasma melatonin (MEL) levels, which in turn regulates hepatic insulin-like growth factor I (IGF-I) secretion via the Mel1c receptor. However, the intracellular signaling pathway initiated by Mel1c remains unclear. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service