Skip to Content
Merck
All Photos(1)

Key Documents

SAB3500380

Sigma-Aldrich

Anti-LXR-B antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-LXRB, Anti-Liver X receptor B, Anti-NR1H2, Anti-Nuclear receptor subfamily 1 group H member 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NR1H2(7376)

General description

Liver X receptor β (LXRβ ) is a member of LXRs that belong to the nuclear receptor superfamily. It possesses a DNA-binding domain that requires the ligand-binding domain to heterodimerize with a retinoid X receptor partner for its DNA-binding activity. The protein is expressed in many tissues and the LXRβ gene is localized on human chromosome 19q13.3.

Immunogen

LXR-B antibody was raised against a 14 amino acid peptide from near the amino terminus human LXR-B.

Biochem/physiol Actions

Liver X receptor β (LXRβ) has a role in cell death and cholesterol metabolism. LXRs have been found to regulate inflammatory response in macrophages. They are also involved in the regulation of cell proliferation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500380.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liver x receptors in atherosclerosis and inflammation.
Im SS and Osborne TF
Circulation Research (2011)
A common polymorphism in NR1H2 (LXRbeta) is associated with preeclampsia.
Mouzat K
BMC Medical Genetics (2011)
Liver X receptor ? activation induces pyroptosis of human and murine colon cancer cells.
Derangere V
Cell Death and Differentiation (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service