Skip to Content
Merck
All Photos(1)

Key Documents

SAB2500586

Sigma-Aldrich

Anti-LAT1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LAT(27040)

General description

Linker for activation of T1 (LAT1) is a tyrosine-phosphorylated integral membrane protein. It is encoded by the gene mapped to human chromosome 16p11.2. LAT is found in peripheral blood lymphocytes, spleen, and thymus. In addition, its expression is also observed in various blood cell types including platelets, megakaryocytes, mast cells, and natural killer cells.

Immunogen

Peptide with sequence C-GAPDYENLQELN from the C Terminus of the protein sequence according to NP_055202.1; NP_001014987.1; NP_001014989.1; NP_001014988.1.

Biochem/physiol Actions

Linker for activation of T1 (LAT1) protein is a transmembrane adaptor signaling molecule. It aids in connecting T cell antigen receptor complex (TCR) to intracellular signaling cascades, which is essential for T-cell development and function. Mutation in the LAT1 gene causes severe combined immunodeficiency (SCID).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiara Bacchelli et al.
The Journal of allergy and clinical immunology, 139(2), 634-642 (2016-08-16)
Signaling through the T-cell receptor (TCR) is critical for T-cell development and function. Linker for activation of T cells (LAT) is a transmembrane adaptor signaling molecule that is part of the TCR complex and essential for T-cell development, as demonstrated by
C Windpassinger et al.
Cytogenetic and genome research, 97(3-4), 155-157 (2002-11-20)
We have mapped the LAT gene by radiation hybrid mapping and fluorescence in situ hybridization to chromosome 16p11.2. The complete cDNA sequence of LAT was generated using assembled sequences of cDNA fragments already available. BLAST analysis using the cDNA sequence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service