Skip to Content
Merck
All Photos(1)

Key Documents

S5192

Sigma-Aldrich

SB 222200

≥98% (HPLC), solid

Synonym(s):

(S)-3-Methyl-2-phenyl-N-(1-phenylpropyl)-4-quinolinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H24N2O
CAS Number:
Molecular Weight:
380.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

light yellow

solubility

DMSO: soluble ~28 mg/mL
H2O: insoluble

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

CC[C@H](NC(=O)c1c(C)c(nc2ccccc12)-c3ccccc3)c4ccccc4

InChI

1S/C26H24N2O/c1-3-22(19-12-6-4-7-13-19)28-26(29)24-18(2)25(20-14-8-5-9-15-20)27-23-17-11-10-16-21(23)24/h4-17,22H,3H2,1-2H3,(H,28,29)/t22-/m0/s1

InChI key

MQNYRKWJSMQECI-QFIPXVFZSA-N

Gene Information

Application

The pharmacodynamics profile of SB 222200 enables its use as a tool to study physiological and pathophysiological roles of NK-3 receptor in CNS-modulated behaviors.

Biochem/physiol Actions

Non-peptide NK3 tachykinin receptor antagonist.
SB 222200 is a 2-phenyl-4-quinolinecarboxamides and a selective, reversible and competitive antagonist of human NK-3 receptor that effectively crosses the blood-brain barrier. It inhibits the NK-3 receptor-induced miosis or pupil constriction in conscious rabbits.1,2

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

Sold for research purposes under agreement from Glaxo­Smith­Kline

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A D Medhurst et al.
British journal of pharmacology, 122(3), 469-476 (1997-11-14)
1. Inhibition of NK3 receptor agonist-induced contraction in the rabbit isolated iris sphincter muscle was used to assess the in vitro functional activity of three 2-phenyl-4-quinolinecarboxamides, members of a novel class of potent and selective non-peptide NK3 receptor antagonists. In
J García-Ortega et al.
Human reproduction (Oxford, England), 29(12), 2736-2746 (2014-10-16)
Are neurokinin B (NKB), NK3 receptor (NK3R), kisspeptin (KISS1) and kisspeptin receptor (KISS1R) expressed in human ovarian granulosa cells? The NKB/NK3R and kisspeptin/KISS1R systems are co-expressed and functionally active in ovarian granulosa cells. The NKB/NK3R and KISS1/KISS1R systems are essential
H M Sarau et al.
The Journal of pharmacology and experimental therapeutics, 295(1), 373-381 (2000-09-19)
The pharmacological and pharmacokinetic profile of SB-222200 [(S)-(-)-N-(alpha-ethylbenzyl)-3-methyl-2-phenylquinoline-4-car boxami de], a human NK-3 receptor (hNK-3R) antagonist, was determined. SB-222200 inhibited (125)I-[MePhe(7)]neurokinin B (NKB) binding to Chinese hamster ovary (CHO) cell membranes stably expressing the hNK-3 receptor (CHO-hNK-3R) with a K(i)
Xinhuai Liu et al.
eLife, 10 (2021-01-20)
The necessity and functional significance of neurotransmitter co-transmission remains unclear. The glutamatergic 'KNDy' neurons co-express kisspeptin, neurokinin B (NKB), and dynorphin and exhibit a highly stereotyped synchronized behavior that reads out to the gonadotropin-releasing hormone (GnRH) neuron dendrons to drive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service