RAB0217
Human Heat Shock Protein 27 ELISA Kit
for serum, plasma, cell culture supernatant and urine
Synonym(s):
HSP27
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type urine
sample type plasma
sample type serum
sample type cell culture supernatant(s)
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 150 pg/mL
standard curve range: 0.205-50 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... HSPB1(3315)
General description
The Human HSP27 (Heat Shock Protein 70) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human HSP27 in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Human HSP27
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8B - Non-combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Akhilesh Kumar et al.
Cell stress & chaperones, 25(3), 509-517 (2020-03-25)
The present study was attempted to investigate the dynamics of HSPA1A and redox status in the spermatozoa and fluid of different segments of buck epididymis. Testes were collected from sexually mature and healthy bucks aged between 2 and 3 years. The
Effect of light intensity on survival, growth and physiology of rohu, Labeo rohita (Cyprinidae) fry.
Nawaz Alam Khan et al.
International journal of radiation biology, 96(4), 552-559 (2019-12-20)
Purpose: Light intensity is one of the important environmental factors that have strong influence on larviculture. Culture of fish at high intensity of light generates harmful radicals in the body that can compromise their health and production. The present study
Biqiong Ren et al.
Oncology reports, 40(2), 895-901 (2018-06-15)
Curcumin has been revealed to inhibit liver cancer, however, no studies have reported that the mechanism of curcumin's action on liver cancer is related to damage-associated molecular pattern (DAMP) molecules heat shock protein 70 (HSP70) and the toll-like receptor 4 (TLR4)
Joeri Lambrecht et al.
EBioMedicine, 43, 501-512 (2019-05-01)
Platelet Derived Growth Factor Receptor beta (PDGFRβ) has been associated to hepatic stellate cell activation and has been the target of multiple therapeutic studies. However, little is known concerning its use as a diagnostic agent. Circulating PDGFRβ levels were analysed
Jin Song et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 28(1), 174-182 (2018-10-20)
Pancreatic cancer is a deadly disease for which available biomarkers, such as CA19-9, lack the desired sensitivity and specificity for early detection. Additional biomarkers are needed to improve both its sensitivity and specificity. Multiplex immunoassays were developed for selected biomarkers
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service