Skip to Content
Merck
All Photos(1)

Key Documents

PZ0407

Sigma-Aldrich

PF-04957325

≥98% (HPLC)

Synonym(s):

3-[[(2R)-4-(2-Thiazolylmethyl)-2-morpholinyl]methyl]-5-(trifluoromethyl)-3H-1,2,3-triazolo[4,5-d]pyrimidin-7-amine, 3-[[(2R)-4-(thiazol-2-ylmethyl)morpholin-2-yl]methyl]-5-(trifluoromethyl)triazolo[4,5-d]pyrimidin-7-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15F3N8OS
CAS Number:
Molecular Weight:
400.38
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(C1=NC2=C(C(N)=N1)N=NN2C[C@@H]3OCCN(C3)CC4=NC=CS4)F

InChI

1S/C14H15F3N8OS/c15-14(16,17)13-20-11(18)10-12(21-13)25(23-22-10)6-8-5-24(2-3-26-8)7-9-19-1-4-27-9/h1,4,8H,2-3,5-7H2,(H2,18,20,21)/t8-/m1/s1

InChI key

VNLPYEMGHGDRRZ-MRVPVSSYSA-N

Biochem/physiol Actions

PF-04957325 is a cAMP-competitive, potent and selective phosphodiesterase 8 inhibitor (IC50 = 3.05 nM/PDE8A, 0.44 nM/PDE8B, 1 μM/PDE4D with 12-14 nM cAMP; >1 μM/all other PDEs).

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service