Skip to Content
Merck
All Photos(1)

Documents

PZ0197

Sigma-Aldrich

Pelitinib

≥98% (HPLC)

Synonym(s):

(2E)-N-[4-[(3-Chloro-4-fluorophenyl)amino]-3-cyano-7-ethoxy-6-quinolinyl]-4-(dimethylamino)-2-butenamide, EKB 569, EKB-569, WAY-EKB-569

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H23ClFN5O2
CAS Number:
Molecular Weight:
467.92
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated
protect from light

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

InChI

1S/C24H23ClFN5O2/c1-4-33-22-12-20-17(11-21(22)30-23(32)6-5-9-31(2)3)24(15(13-27)14-28-20)29-16-7-8-19(26)18(25)10-16/h5-8,10-12,14H,4,9H2,1-3H3,(H,28,29)(H,30,32)/b6-5+

InChI key

WVUNYSQLFKLYNI-AATRIKPKSA-N

Biochem/physiol Actions

Pelitinib (EKB-569) is an orally active, potent, second generation irreversible epidermal growth factor receptor tyrosine kinase (EGFR TK) inhibitor. Pelitinib is selective for EGFR (ErbB-1) with an IC50 about 80 nM, but also covalently binds to ErbB-2 (HER2/c-neu) and ErbB-4 (Her 4) with much lower potency. Pelitinib has been shown to inhibit the growth of EGFR-overexpressing tumor cell lines.
Pelitinib specifically targets cancer cells with elevated expression of the efflux transporters ATP-binding cassette (ABC)B1/ABCG2 post hyperthermia. This property of peltinib aids in the elimination of metastasis and prevention of cancer recurrence.

Other Notes

Light sensitive

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molly F Kulesz-Martin et al.
Cancer biology & therapy, 14(2), 95-99 (2012-11-30)
Current therapies for Renal Cell Carcinoma favor vascular endothelial growth factor receptor (VEGF-R) tyrosine kinase (TK) inhibitors (TKIs). In theory, these are most applicable in tumors that have lost VHL-with subsequent stabilization of HIF and upregulation of VEGF. A subset
Shan Bian et al.
Nature methods, 15(8), 631-639 (2018-07-25)
Brain tumors are among the most lethal and devastating cancers. Their study is limited by genetic heterogeneity and the incompleteness of available laboratory models. Three-dimensional organoid culture models offer innovative possibilities for the modeling of human disease. Here we establish
Kenneth K W To et al.
British journal of pharmacology, 172(16), 4089-4106 (2015-05-20)
Pelitinib is a potent irreversible EGFR TK inhibitor currently in clinical trials for the treatment of lung cancer. Hyperthermia has been applied concomitantly with chemotherapy and radiotherapy to enhance treatment outcome. In this study, we investigated the ability of the
Yeong Hoon Kim et al.
The Korean journal of parasitology, 55(5), 491-503 (2017-11-07)
The effects of tyrosine kinase inhibitors (TKIs) were evaluated on growth inhibition of intracellular Toxoplasma gondii in host ARPE-19 cells. The number of tachyzoites per parasitophorous vacuolar membrane (PVM) was counted after treatment with TKIs. T. gondii protein expression was
Jean-Philippe Coppé et al.
Nature cell biology, 21(6), 778-790 (2019-06-05)
Phosphorylation networks intimately regulate mechanisms of response to therapies. Mapping the phospho-catalytic profile of kinases in cells or tissues remains a challenge. Here, we introduce a practical high-throughput system to measure the enzymatic activity of kinases using biological peptide targets

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service