Skip to Content
Merck
All Photos(1)

Documents

PLA0178

Sigma-Aldrich

Rabbit anti-p16INK4a Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

ARF, CDK4 inhibitor p16-INK4, CDK4I, CDKN2, CMM2, INK4, INK4a, MLM, MTS-1, MTS1, TP16, alternative reading frame, cell cycle negative regulator beta, cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A (melanoma; p16; inhibits CDK4), multiple tumor suppressor 1, p14, p14ARF, p16, p16-INK4, p16INK4a, p19, p19Arf

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

concentration

0.2 mg/mL

technique(s)

immunohistochemistry: 1:100-1:500
immunoprecipitation (IP): 2-10 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

AAA92554.1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... p16INK4a(1029)

General description

Cyclin-dependent kinase inhibitor 2A (CDK4I) or p16INK4a belongs to the Ink4 family of CDK inhibitors. The p16INK4agene is mapped to human chromosome 9p21. Structurally, p16INK4a comprises four ankyrin-type repeats (I–IV) .

Immunogen

The epitope recognized by PLA0178 maps to a region between residue 106 and 156 of human CDK4 inhibitor p16-INK4a using the numbering given in entry AAA92554.1 (GeneID 1029).

Biochem/physiol Actions

Cyclin-dependent kinase inhibitor 2A (CDK4I) or p16INK4a is a negative regulator of cyclin dependant kinase 4 (CDK4) and CDK6 kinase activity and cell cycle progression. The ankyrin repeats (I–IV) are crucial for their interaction with CDK4 and CDK6. P16INK4a regulates the cell cycle by inhibiting cyclin/CDK complex formation and the phosphorylation of the retinoblastoma protein (pRb). P16INK4a is a tumor suppressor and its mutations are associated with malignancies such as melanoma and astrocytoma and are the cause of Li-Fraumeni syndrome.

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

p16INK4a is a negative regulator of CDK4 and CDK6 kinase activity and cell cycle progression. P16INK4a regulates the cell cycle by inhibiting cyclin/CDK complex formation and the phosphorylation of the retinoblastoma protein (pRb). P16INK4a is a tumor suppressor associated with malignancies such as melanoma and astrocytoma and is the cause of Li-Fraumeni syndrome.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sock Hoai Chan et al.
NPJ genomic medicine, 1, 16015-16015 (2016-06-01)
Li-Fraumeni syndrome (LFS) is a rare cancer predisposition syndrome usually associated with TP53 germline alterations. Its genetic basis in TP53 wild-type pedigrees is less understood. Using whole-genome sequencing, we identified a germline hemizygous deletion ablating CDKN2A-CDKN2B in a TP53 wild-type
Caroline Kannengiesser et al.
Human mutation, 30(4), 564-574 (2009-03-05)
Germline mutations of the CDKN2A gene are found in melanoma-prone families and individuals with multiple sporadic melanomas. The encoded protein, p16(INK4A), comprises four ankyrin-type repeats, and the mutations, most of which are missense and occur throughout the entire coding region
p16INK4a: a central player in cellular senescence and a promising aging biomarker in elderly cancer patients
Vandenberk B, et al.
2009 IEEE 3rd International Conference on Nano/Molecular Medicine and Engineering, 2(4) (2011)
David Machover et al.
The Journal of pharmacology and experimental therapeutics, 369(3), 489-502 (2019-04-04)
Methionine deprivation induces growth arrest and death of cancer cells. To eliminate l-methionine we produced, purified, and characterized the recombinant pyridoxal 5'-phosphate (PLP)-dependent l-methionine γ-lyase (MGL)- BL929 from the cheese-ripening Brevibacterium aurantiacum Transformation of an Escherichia coli strain with the
C Romagosa et al.
Oncogene, 30(18), 2087-2097 (2011-02-08)
p16(Ink4a) is a protein involved in regulation of the cell cycle. Currently, p16(Ink4a) is considered a tumor suppressor protein because of its physiological role and downregulated expression in a large number of tumors. Intriguingly, overexpression of p16(Ink4a) has also been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service